accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76067-81-7

IUPAC Name: 2-(3,5-diaminobenzoyloxy)ethyl 2-methylprop-2-enoate
Synonyms: 2-(3,5-diaminobenzoyloxy)ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 264.28
Molecular Formula: C13H16N2O4
InChi Key: BZRAULNYWZRKMB-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOC(=O)C1=CC(N)=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results