Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methacryloyloxy)ethyl 3,5-diaminobenzoate, 99%, Thermo Scientific™

Catalog No. AAH2584003
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2584003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2584003 Supplier Thermo Scientific Chemicals Supplier No. H2584003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 76067-81-7
Molecular Formula C13H16N2O4
Quantity 1g
MDL Number MFCD00425081
Synonym 2-methacryloyloxy ethyl 3,5-diaminobenzoate, 2-3,5-diaminobenzoyloxy ethyl 2-methylprop-2-enoate, 2-2-methylprop-2-enoyloxy ethyl 3,5-diaminobenzoate, methacrylic acid 2-3,5-diaminobenzoyloxy ethyl ester, 3,5-diaminobenzoic acid 2-methacryloyloxy ethyl ester, benzoic acid,3,5-diamino-, 2-2-methyl-1-oxo-2-propen-1-yl oxy ethyl ester
InChI Key BZRAULNYWZRKMB-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OCCOC(=O)C1=CC(N)=CC(N)=C1
IUPAC Name 2-(3,5-diaminobenzoyloxy)ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 264.28
PubChem CID 2306460
Formula Weight 264.28
Percent Purity 99%
Chemical Name or Material 2-(Methacryloyloxy)ethyl 3,5-diaminobenzoate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.