accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSOOHNNNClCl

CAS RN 761440-16-8

IUPAC Name: 2,5-dichloro-N-[2-(propane-2-sulfonyl)phenyl]pyrimidin-4-amine
Synonyms: 2,5-dichloro-N-[2-(propane-2-sulfonyl)phenyl]pyrimidin-4-amine
Molecular Weight (g/mol): 346.23
Molecular Formula: C13H13Cl2N3O2S
InChi Key: WWVLDJAVSQZSKO-UHFFFAOYSA-N
SMILES: CC(C)S(=O)(=O)C1=CC=CC=C1NC1=NC(Cl)=NC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results