accessibility menu, dialog, popup

UserName

Viewing profile as user:
BClOCH3HOOH

CAS RN 762287-57-0

IUPAC Name: (4-chloro-2-methoxyphenyl)boronic acid
Synonyms: 4-chloro-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 186.4
Molecular Formula: C7H8BClO3
InChi Key: NZRRMTBNTSBIFH-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(Cl)=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results