CAS RN 7627-80-7
IUPAC Name:
tetrafluoropyridazine
Synonyms:
tetrafluoropyridazine
Molecular Weight (g/mol):
152.05
Molecular Formula:
C4F4N2
InChi Key:
WZGAMTYFRMRZNL-UHFFFAOYSA-N
SMILES:
FC1=NN=C(F)C(F)=C1F
1
–
1
of
1
results
1
Tetrafluoropyridazine, 97%, Thermo Scientific™
CAS: 7627-80-7 Molecular Formula: C4F4N2 Molecular Weight (g/mol): 152.052 MDL Number: MFCD04972969 InChI Key: WZGAMTYFRMRZNL-UHFFFAOYSA-N Synonym: pyridazine, tetrafluoro,tetrafluoropyridazine,pyridazine,tetrafluoro,perfluoropyridazine,3,4,5,6-tetrafluoro-pyridazine PubChem CID: 139055 IUPAC Name: 3,4,5,6-tetrafluoropyridazine SMILES: C1(=C(C(=NN=C1F)F)F)F