Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrafluoropyridazine, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
For preparing perfluoro-aromatic nitrogen heterocyclic compounds from their perchloro-analogues by exchange with potassium fluoride in the absence of solvent has been extended to the pyridazine system. Tetrafluoropyridazine readily
For preparing perfluoro-aromatic nitrogen heterocyclic compounds from their perchloro-analogues by exchange with potassium fluoride in the absence of solvent has been extended to the pyridazine system. Tetrafluoropyridazine readily
Specifications
Specifications
| Boiling Point | 117°C (160 mmHg) |
| Quantity | 500 mg |
| Formula Weight | 152.05 |
| Percent Purity | 97% |
| Chemical Name or Material | Tetrafluoropyridazine |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P301+P310-P305+P351+P338-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?