accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 763-69-9

IUPAC Name: ethyl 3-ethoxypropanoate
Synonyms: ethyl 3-ethoxypropanoate
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: BHXIWUJLHYHGSJ-UHFFFAOYSA-N
SMILES: CCOCCC(=O)OCC
Boiling Point 
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)