accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHNHN

CAS RN 763120-58-7

IUPAC Name: (1H-pyrazol-4-yl)boronic acid
Synonyms: 1H-pyrazol-4-ylboronic acid
Molecular Weight (g/mol): 111.9
Molecular Formula: C3H5BN2O2
InChi Key: KEZNMOUMHOZFRA-UHFFFAOYSA-N
SMILES: OB(O)C1=CNN=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results