accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrH2C

CAS RN 76334-36-6

IUPAC Name: 3-bromobut-3-en-1-ol
Synonyms: 3-bromo-3-buten-1-ol C4H7BrO
Molecular Weight (g/mol): 151
Molecular Formula: C4H7BrO
InChi Key: RTKMFQOHBDVEBC-UHFFFAOYSA-N
SMILES: OCCC(Br)=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)