accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7635-54-3

IUPAC Name: (1S,2R,5S)-5-methyl-2-(propan-2-yl)cyclohexyl carbonochloridate
Synonyms: (1S,2R,5S)-2-isopropyl-5-methylcyclohexyl carbonochloridate
Molecular Weight (g/mol): 218.72
Molecular Formula: C11H19ClO2
InChi Key: KIUPCUCGVCGPPA-AEJSXWLSSA-N
SMILES: CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results