accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76350-90-8

IUPAC Name: {2-methyl-[1,1'-biphenyl]-3-yl}methanol
Synonyms: {2-methyl-[1,1'-biphenyl]-3-yl}methanol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: BGTLHJPGBIVQLJ-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=CC=C1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results