Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxymethyl-2-methylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. H077725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H077725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H077725G Supplier TCI America Supplier No. H077725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76350-90-8
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00134200
InChI Key BGTLHJPGBIVQLJ-UHFFFAOYSA-N
Synonym 2-Methylbiphenyl-3-methanol
PubChem CID 596875
IUPAC Name (2-methyl-3-phenylphenyl)methanol
SMILES CC1=C(C=CC=C1CO)C2=CC=CC=C2

Specifications

Melting Point 72°C
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxymethyl-2-methylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.