accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 76357-18-1

IUPAC Name: methyl 1-(triphenylmethyl)aziridine-2-carboxylate
Synonyms: methyl 1-(triphenylmethyl)aziridine-2-carboxylate
Molecular Weight (g/mol): 343.43
Molecular Formula: C23H21NO2
InChi Key: QGSITPKXYQEFIR-UHFFFAOYNA-N
SMILES: COC(=O)C1CN1C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results