Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-(Triphenylmethyl)-2-aziridinecarboxylate, TCI America™
SDP

Catalog No. M19275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19275G Supplier TCI America Supplier No. M19275G

Chemical Identifiers

CAS 76357-18-1
Molecular Formula C23H21NO2
Molecular Weight (g/mol) 343.426
InChI Key QGSITPKXYQEFIR-UHFFFAOYSA-N
Synonym Methyl 1-Trityl-2-aziridinecarboxylate, 1-Trityl-2-aziridinecarboxylic Acid Methyl Ester
PubChem CID 291744
IUPAC Name methyl 1-tritylaziridine-2-carboxylate
SMILES COC(=O)C1CN1C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Formula Weight 343.43
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-(Triphenylmethyl)-2-aziridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.