accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7646-67-5

IUPAC Name: N-(2-hydroxyethyl)prop-2-enamide
Synonyms: N-(2-hydroxyethyl)prop-2-enamide
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: UUORTJUPDJJXST-UHFFFAOYSA-N
SMILES: OCCNC(=O)C=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)