Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Hydroxyethyl acrylamide

Catalog No. 697931100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-793-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-793-1100ML Supplier Sigma Aldrich Supplier No. 697931100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Hydroxyethyl acrylamide is an amphipathic polymer that can be used for a variety of applications such as:preparation of hydrogels for the formation of a stimulo-responsive polymer for cell signaling and bioengineering researchin the synthesis of lanthanide based luminescent materials for sensors and bio-imagingformation of adhesive hydrogel for potential usage in the smart adhesives

Specifications

CAS 7646-67-5
Density 1.111 g/mL (at 25°C)
Boiling Point 129°C to 130°C
Molecular Formula C5H9NO2
Refractive Index n20/D 1.505
Linear Formula C5H9NO2
Quantity 100 mL
MDL Number MFCD11110645
Synonym N-(2-Hydroxyethyl)acrylamide; HEAA
Molecular Weight (g/mol) 115.13
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.