CAS RN 7647-18-9
Antimony(V) chloride, ultra dry, 99.999% (metals basis)
CAS: 7647-18-9 Molecular Formula: Cl5Sb Molecular Weight (g/mol): 299.02 MDL Number: MFCD00011213 InChI Key: VMPVEPPRYRXYNP-UHFFFAOYSA-I Synonym: antimony v chloride,antimony pentachloride,pentachloroantimony,antimony perchloride,antimony chloride sbcl5,antimonpentachlorid,antimoonpentachloride,perchlorure d'antimoine,pentachlorure d'antimoine,antimonpentachlorid german PubChem CID: 24294 IUPAC Name: antimony pentachloride SMILES: Cl[Sb](Cl)(Cl)(Cl)Cl
Antimony(V) chloride, 99.997% (metals basis)
CAS: 7647-18-9 Molecular Formula: Cl5Sb Molecular Weight (g/mol): 299.02 MDL Number: MFCD00011213 InChI Key: VMPVEPPRYRXYNP-UHFFFAOYSA-I Synonym: antimony v chloride,antimony pentachloride,pentachloroantimony,antimony perchloride,antimony chloride sbcl5,antimonpentachlorid,antimoonpentachloride,perchlorure d'antimoine,pentachlorure d'antimoine,antimonpentachlorid german PubChem CID: 24294 IUPAC Name: antimony pentachloride SMILES: Cl[Sb](Cl)(Cl)(Cl)Cl
Antimony(V)-chloride, 99%, anhydrous
CAS: 7647-18-9 Molecular Formula: Cl5Sb Molecular Weight (g/mol): 299.02 MDL Number: MFCD00011213 InChI Key: VMPVEPPRYRXYNP-UHFFFAOYSA-I Synonym: antimony v chloride,antimony pentachloride,pentachloroantimony,antimony perchloride,antimony chloride sbcl5,antimonpentachlorid,antimoonpentachloride,perchlorure d'antimoine,pentachlorure d'antimoine,antimonpentachlorid german PubChem CID: 24294 IUPAC Name: pentachloro-$l^{5}-stibane SMILES: Cl[Sb](Cl)(Cl)(Cl)Cl
Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
CAS: 7647-18-9 | Molecular Formula: Cl5Sb | Molecular Weight: 299.02 g/mol
Sigma Aldrich Antimony(V) chloride solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Antimony(V) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Antimony(V) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Antimony(V) chloride, 98%
CAS: 7647-18-9 Molecular Formula: Cl5Sb Molecular Weight (g/mol): 299.02 MDL Number: MFCD00011213 InChI Key: VMPVEPPRYRXYNP-UHFFFAOYSA-I Synonym: antimony v chloride,antimony pentachloride,pentachloroantimony,antimony perchloride,antimony chloride sbcl5,antimonpentachlorid,antimoonpentachloride,perchlorure d'antimoine,pentachlorure d'antimoine,antimonpentachlorid german PubChem CID: 24294 IUPAC Name: antimony pentachloride SMILES: Cl[Sb](Cl)(Cl)(Cl)Cl