Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(V) chloride, 98%

Catalog No. AA41978A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41978A1 1 kg
AA4197818 50 g
AA4197830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41978A1 Supplier Thermo Scientific Chemicals Supplier No. 41978A1

missing translation for 'validMainframeMsg'

CAS: 7647-18-9 | Cl5Sb | 299.02 g/mol

Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name antimony pentachloride
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 79°C (22 mm)
Melting Point 2.8°C
Color Red-Yellow
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 98%
Solubility Information Soluble in HCl,chloroform,carbon tetrachlorideMiscible with chloroform,hydrochloric acid,tartaric acid,carbon disulfide and carbon tetrachloride.
Formula Weight 299.02
Odor Pungent
Refractive Index 1.601
Sensitivity Moisture sensitive
Appearance Red-yellow liquid
Density 2.336
Chemical Name or Material Antimony(V) chloride
Show More Show Less
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-601-8
RTECSNumber CC5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.