accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONH(R)NHOOOHO

CAS RN 76477-42-4

IUPAC Name: (2R)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2R)-6-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 380.44
Molecular Formula: C19H28N2O6
InChi Key: BDHUTRNYBGWPBL-GGYSOQFKNA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results