accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)SH3COOO

CAS RN 76497-39-7

IUPAC Name: (2S)-3-(acetylsulfanyl)-2-methylpropanoate
Synonyms: (2S)-3-(acetylsulfanyl)-2-methylpropanoate
Molecular Weight (g/mol): 161.2
Molecular Formula: C6H9O3S
InChi Key: VFVHNRJEYQGRGE-SCSAIBSYSA-M
SMILES: C[C@H](CSC(C)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results