Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-(Acetylthio)-2-methylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. A133425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A133425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A133425ML Supplier TCI America Supplier No. A133425ML

Specifications

CAS 76497-39-7
Melting Point 5°C
Color Yellow
Boiling Point 132°C
Molecular Formula C6H10O3S
MDL Number MFCD00038563
Quantity 25 mL
InChI Key VFVHNRJEYQGRGE-SCSAIBSYSA-N
SMILES CC(CSC(=O)C)C(=O)O
IUPAC Name (2S)-3-acetylsulfanyl-2-methylpropanoic acid
Molecular Weight (g/mol) 162.203
PubChem CID 2724506
Formula Weight 162.20
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-3-(Acetylthio)-2-methylpropionic Acid
Show More Show Less
EINECSNumber (2)-3239
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.