accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 765-12-8

IUPAC Name: 3,6,9,12-tetraoxatetradeca-1,13-diene
Synonyms: 3,6,9,12-tetraoxatetradeca-1,13-diene
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: CYIGRWUIQAVBFG-UHFFFAOYSA-N
SMILES: C=COCCOCCOCCOC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)