accessibility menu, dialog, popup

UserName

Viewing profile as user:
P

CAS RN 7650-89-7

IUPAC Name: tribenzylphosphane
Synonyms: tribenzylphosphine
Molecular Weight (g/mol): 304.37
Molecular Formula: C21H21P
InChi Key: IFXORIIYQORRMJ-UHFFFAOYSA-N
SMILES: C(P(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)