accessibility menu, dialog, popup

UserName

Viewing profile as user:
P

CAS RN 7650-91-1

IUPAC Name: benzyldiphenylphosphane
Synonyms: benzyldiphenylphosphane
Molecular Weight (g/mol): 276.32
Molecular Formula: C19H17P
InChi Key: UZCPNEBHTFYJNY-UHFFFAOYSA-N
SMILES: C(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results