accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7651-81-2

IUPAC Name: 2,3-dihydroisoquinolin-3-one
Synonyms: 2H-isoquinolin-3-one
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: GYPOFOQUZZUVQL-UHFFFAOYSA-N
SMILES: O=C1NC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)