Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyisoquinoline 98.0+%, TCI America™
SDP

Catalog No. H14515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14515G 5 g
H14511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14515G Supplier TCI America Supplier No. H14515G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7651-81-2
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00075524
InChI Key GYPOFOQUZZUVQL-UHFFFAOYSA-N
Synonym 3-hydroxyisoquinoline, isoquinolin-3-ol, isoquinolin-3 2h-one, 3-isoquinolinol, 2,3-dihydroisoquinolin-3-one, 3 2h-isoquinolinone, 2h-3-isoquinolone, zlchem 163, 3-hydroxy-isoquinoline, 3-isoquinolinol 9ci
PubChem CID 2736554
IUPAC Name 2H-isoquinolin-3-one
SMILES C1=CC2=CC(=O)NC=C2C=C1

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.