accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 765317-72-4

IUPAC Name: 3-(3,5-dibromo-4-hydroxybenzoyl)-2-ethyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}-1-benzofuran-6-sulfonamide
Synonyms: 3-(3,5-dibromo-4-hydroxybenzoyl)-2-ethyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}-1-benzofuran-6-sulfonamide
Molecular Weight (g/mol): 741.44
Molecular Formula: C26H19Br2N3O7S3
InChi Key: SXKBTDJJEQQEGE-UHFFFAOYSA-N
SMILES: CCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=C(C=C2O1)S(=O)(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results