accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3

CAS RN 7654-51-5

IUPAC Name: 4,4'-dimethyl-2,2'-biquinoline
Synonyms: 4,4'-dimethyl-2,2'-biquinoline
Molecular Weight (g/mol): 284.36
Molecular Formula: C20H16N2
InChi Key: DXZWINPECYHJIG-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC(=C1)C1=CC(C)=C2C=CC=CC2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results