Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bi-4-lepidine 98.0+%, TCI America™
SDP

Catalog No. B0841100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0841100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0841100MG Supplier TCI America Supplier No. B0841100MG

Chemical Identifiers

CAS 7654-51-5
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.36
MDL Number MFCD00059753
InChI Key DXZWINPECYHJIG-UHFFFAOYSA-N
Synonym 2,2′C-Di-4-lepidyl, 4,4′C-Dimethyl-2,2′C-biquinoline
PubChem CID 2827924
IUPAC Name 4,4'-dimethyl-2,2'-biquinoline
SMILES CC1=C2C=CC=CC2=NC(=C1)C1=CC(C)=C2C=CC=CC2=N1

Specifications

Melting Point 281°C
Quantity 100 mg
Formula Weight 284.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bi-4-lepidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.