CAS RN 76541-44-1
IUPAC Name:
3-(difluoromethyl)pyridine
Synonyms:
3-(difluoromethyl)pyridine
Molecular Weight (g/mol):
129.11
Molecular Formula:
C6H5F2N
InChi Key:
HKRKGTFBASUFRO-UHFFFAOYSA-N
SMILES:
FC(F)C1=CC=CN=C1
1
–
2
of
2
results
1
Promotions Available
3-(Difluoromethyl)pyridine, 97%
3-(Difluoromethyl)pyridine, 97%
CAS: 76541-44-1 Molecular Formula: C6H5F2N Molecular Weight (g/mol): 129.11 MDL Number: MFCD11226584 InChI Key: HKRKGTFBASUFRO-UHFFFAOYSA-N Synonym: 3-difluoromethyl pyridine,pyridine, 3-difluoromethyl,pyridine,3-difluoromethyl,pyridine, 3-difluoromethyl-9ci PubChem CID: 640475 IUPAC Name: 3-(difluoromethyl)pyridine SMILES: FC(F)C1=CC=CN=C1
2
Sigma Aldrich 3-(Difluoromethyl)pyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.