accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClN

CAS RN 7659-45-2

IUPAC Name: 3-chloro-2-methylpropanenitrile
Synonyms: 3-chloro-2-methylpropanenitrile
Molecular Weight (g/mol): 103.55
Molecular Formula: C4H6ClN
InChi Key: XXZVBSPSFVERCB-UHFFFAOYNA-N
SMILES: CC(CCl)C#N
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results