CAS RN 766-66-5
CAS RN 766-66-5
IUPAC Name:
2-chlorocycloheptan-1-one
Synonyms:
cycloheptanone, 2-chloro-
Molecular Weight (g/mol):
146.61
Molecular Formula:
C7H11ClO
InChi Key:
SDSZGDVXUFWFAA-UHFFFAOYNA-N
SMILES:
ClC1CCCCCC1=O
1
–
1
of
1
results
1
2-Chlorocycloheptanone 98.0+%, TCI America™
CAS: 766-66-5 Molecular Formula: C7H11ClO Molecular Weight (g/mol): 146.61 MDL Number: MFCD00041033 InChI Key: SDSZGDVXUFWFAA-UHFFFAOYNA-N PubChem CID: 13012 IUPAC Name: 2-chlorocycloheptan-1-one SMILES: ClC1CCCCCC1=O