Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorocycloheptanone 98.0+%, TCI America™
SDP

Catalog No. C16455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16455G Supplier TCI America Supplier No. C16455G

Chemical Identifiers

CAS 766-66-5
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00041033
InChI Key SDSZGDVXUFWFAA-UHFFFAOYNA-N
PubChem CID 13012
IUPAC Name 2-chlorocycloheptan-1-one
SMILES ClC1CCCCCC1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 146.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorocycloheptanone
RTECSNumber GU3500000
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.