accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7664-80-4

H3COOSH

CAS RN 7664-80-4

IUPAC Name: octyl 2-sulfanylacetate
Synonyms: octyl 2-sulfanylacetate
Molecular Weight (g/mol): 204.33
Molecular Formula: C10H20O2S
InChi Key: MADOXCFISYCULS-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results