Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl Thioglycolate 95.0+%, TCI America™
SDP

Catalog No. T062725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T062725G Supplier TCI America Supplier No. T062725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-80-4
Molecular Formula C10H20O2S
Molecular Weight (g/mol) 204.328
MDL Number MFCD00053518
InChI Key MADOXCFISYCULS-UHFFFAOYSA-N
Synonym Mercaptoacetic Acid n-Octyl Ester, n-Octyl Mercaptoacetate, Thioglycolic Acid n-Octyl Ester
PubChem CID 76023
IUPAC Name octyl 2-sulfanylacetate
SMILES CCCCCCCCOC(=O)CS

Specifications

Color Colorless
Boiling Point 124°C
Quantity 25 g
UN Number 2810
Formula Weight 204.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Thioglycolate
EINECSNumber (2)-1359
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.