CAS RN 7673-09-8
IUPAC Name:
N-[4,6-bis(chloroamino)-1,3,5-triazin-2-yl]chloranamine
Synonyms:
trichloro melamine
Molecular Weight (g/mol):
229.45
Molecular Formula:
C3H3Cl3N6
InChi Key:
KEPNSIARSTUPGS-UHFFFAOYSA-N
SMILES:
ClNC1=NC(NCl)=NC(NCl)=N1
1
–
1
of
1
results
1
Trichloromelamine 95.0+%, TCI America™
CAS: 7673-09-8 Molecular Formula: C3H3Cl3N6 Molecular Weight (g/mol): 229.45 MDL Number: MFCD00006047 InChI Key: KEPNSIARSTUPGS-UHFFFAOYSA-N Synonym: 2,4,6-Tris(chloroamino)-1,3,5-triazine PubChem CID: 24322 IUPAC Name: 2-N,4-N,6-N-trichloro-1,3,5-triazine-2,4,6-triamine SMILES: C1(=NC(=NC(=N1)NCl)NCl)NCl