Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloromelamine 95.0+%, TCI America™
SDP

Catalog No. T038425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T038425G 25 g
T0384100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T038425G Supplier TCI America Supplier No. T038425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7673-09-8
Molecular Formula C3H3Cl3N6
Molecular Weight (g/mol) 229.45
MDL Number MFCD00006047
InChI Key KEPNSIARSTUPGS-UHFFFAOYSA-N
Synonym 2,4,6-Tris(chloroamino)-1,3,5-triazine
PubChem CID 24322
IUPAC Name 2-N,4-N,6-N-trichloro-1,3,5-triazine-2,4,6-triamine
SMILES C1(=NC(=NC(=N1)NCl)NCl)NCl

Specifications

Color Yellow
Quantity 25 g
UN Number 1479
Formula Weight 229.45
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trichloromelamine
RTECSNumber XZ1575000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.