accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNHSNOOHN

CAS RN 7675-46-9

IUPAC Name: 2-{[(2-nitrophenyl)sulfanyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-{[(2-nitrophenyl)sulfanyl]amino}propanoic acid; dicha
Molecular Weight (g/mol): 423.57
Molecular Formula: C21H33N3O4S
InChi Key: LGTGUPPUIQQLBP-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(NSC1=CC=CC=C1[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results