accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 768-56-9

IUPAC Name: (but-3-en-1-yl)benzene
Synonyms: benzene, 3-butenyl-
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: PBGVMIDTGGTBFS-UHFFFAOYSA-N
SMILES: C=CCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)