accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNCH3OOHO

CAS RN 7682-16-8

IUPAC Name: 2-acetamidohexanoic acid
Synonyms: D-norleucine, N-acetyl-
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: JDMCEGLQFSOMQH-UHFFFAOYNA-N
SMILES: CCCCC(NC(C)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results