Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-DL-norleucine 98.0+%, TCI America™
SDP

Catalog No. A075025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A075025G 25 g
A07501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A075025G Supplier TCI America Supplier No. A075025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7682-16-8
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
MDL Number MFCD00037272
InChI Key JDMCEGLQFSOMQH-UHFFFAOYSA-N
Synonym Ac-DL-Nle-OH
PubChem CID 306142
IUPAC Name 2-acetamidohexanoic acid
SMILES CCCCC(C(=O)O)NC(=O)C

Specifications

Melting Point 108°C
Color White
Quantity 25 g
Formula Weight 173.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.