CAS RN 76824-99-2
IUPAC Name:
6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Synonyms:
6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Molecular Weight (g/mol):
193.2
Molecular Formula:
C10H11NO3
InChi Key:
CRAGDYRHPWTZJL-UHFFFAOYNA-N
SMILES:
OC(=O)C1CC2=C(CN1)C=CC(O)=C2
1
–
1
of
1
results
1
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™
CAS: 76824-99-2 Molecular Formula: C10H11NO3 Molecular Weight (g/mol): 193.20 MDL Number: MFCD03092912 InChI Key: CRAGDYRHPWTZJL-UHFFFAOYNA-N Synonym: 6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate,6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate,akos 68,d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate,6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o,6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate,d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid PubChem CID: 18525693 SMILES: OC(=O)C1CC2=C(CN1)C=CC(O)=C2