accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76835-17-1

NNHClClHCl

CAS RN 76835-17-1

IUPAC Name: 1-(3,4-dichlorophenyl)piperazine hydrochloride
Synonyms: 1-(3,4-dichlorophenyl)piperazine hydrochloride
Molecular Weight (g/mol): 267.58
Molecular Formula: C10H13Cl3N2
InChi Key: ITTMUMIEDXNQEQ-UHFFFAOYSA-N
SMILES: Cl.ClC1=C(Cl)C=C(C=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results