Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dichlorophenyl)piperazine monohydrochloride, 98+%, Thermo Scientific™

Catalog No. AAB2199509
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2199509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2199509 Supplier Thermo Scientific Chemicals Supplier No. B2199509

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76835-17-1
Molecular Formula C10H13Cl3N2
Molecular Weight (g/mol) 267.578
MDL Number MFCD00190237
InChI Key ITTMUMIEDXNQEQ-UHFFFAOYSA-N
Synonym 1-3,4-dichlorophenyl piperazine hydrochloride, 1-3,4-dichlorophenyl piperazine monohydrochloride, 1-3,4-dichlorophenyl piperazinehydrochloride, pubchem20987, 3,4-dichlorophenyl piperazine, chloride, 1-3,4-dichlorophenyl piperazine hcl, 4-3,4-dichlorophenyl-piperazine hydrochloride
PubChem CID 15574733
IUPAC Name 1-(3,4-dichlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC(=C(C=C2)Cl)Cl.Cl

Specifications

Melting Point 217°C to 220°C
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 267.59
Percent Purity ≥98%
Chemical Name or Material 1-(3,4-Dichlorophenyl)piperazine monohydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.