accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(Z)

CAS RN 7688-21-3

IUPAC Name: (2Z)-hex-2-ene
Synonyms: 2-hexene (Z)
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: RYPKRALMXUUNKS-HYXAFXHYSA-N
SMILES: CCC\C=C/C
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)