Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich cis-2-Hexene

Catalog No. 5384935G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-849-35G 5 g
53-849-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-849-35G Supplier Sigma Aldrich Supplier No. 5384935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cis-2-hexene is one of the isomeric forms of hexene
  • It is obtained from 2-hexyne, via semihydrogenation in the presence of pd78(phen) [phen = 1,10-phenanthroline] on titanium dioxide in n-octane
  • Cis-2-hexene is formed as one of the primary product from the dimerization of propylene in the presence of a nickel oxide-silica-alumina catalyst
  • It undergoes isomerization/hydrosilylation reaction with phenyl silane in the presence of cobalt catalyst to afford 1-hexylphenylsilane

Specifications

CAS 7688-21-3
Density 0.669 g/mL (at 25°C (literature))
Boiling Point 68°C to 70°C (lit.)
Molecular Formula C6H12
Refractive Index n20/D 1.396 (literature)
Linear Formula CH3CH2CH2CH=CHCH3
MDL Number MFCD00066520
Quantity 5 g
Molecular Weight (g/mol) 84.16
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.