accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7693-50-7

IUPAC Name: naphthalen-2-yl carbonochloridate
Synonyms: naphthalen-2-yl carbonochloridate
Molecular Weight (g/mol): 206.63
Molecular Formula: C11H7ClO2
InChi Key: KDUFRLUGTLSJKD-UHFFFAOYSA-N
SMILES: ClC(=O)OC1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results