Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl Chloroformate 97.0+%, TCI America™
SDP

Catalog No. C11155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11155G Supplier TCI America Supplier No. C11155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7693-50-7
Molecular Formula C11H7ClO2
Molecular Weight (g/mol) 206.63
MDL Number MFCD00060059
InChI Key KDUFRLUGTLSJKD-UHFFFAOYSA-N
Synonym Chloroformic Acid 2-Naphthyl Ester
PubChem CID 15764398
IUPAC Name naphthalen-2-yl carbonochloridate
SMILES ClC(=O)OC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 62°C
Color White
Boiling Point 112°C
Quantity 5 g
UN Number 3277
Formula Weight 206.63
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.