accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrNOO

CAS RN 7693-52-9

IUPAC Name: 4-bromo-2-nitrophenol
Synonyms: 4-bromo-2-nitrophenol
Molecular Weight (g/mol): 218.01
Molecular Formula: C6H4BrNO3
InChi Key: CUTFAPGINUFNQM-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)