CAS RN 7699-41-4
CAS RN 7699-41-4
IUPAC Name:
dihydroxysilanone
Synonyms:
dihydroxysilanone
Molecular Weight (g/mol):
78.1
Molecular Formula:
H2O3Si
InChi Key:
IJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILES:
O[Si](O)=O
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich Rhamnolipids, 95% (Mono-Rhamnolipid Dominant)
Sigma Aldrich Rhamnolipids, 95% (Mono-Rhamnolipid Dominant)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.